missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Universal PCR Master Mix, no AmpErase™ UNG

Catalog No. 4324020
Encompass
Change view
Click to view available options
Quantity:
1 x 5 mL, 200 reactions
1 x 50 mL 2,000 reactions
10 x 5 mL, 2,000 reactions
2 x 5 mL, 400 reactions
5 x 5 mL, 1,000 reactions
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4324020 10 x 5 mL, 2,000 reactions
4324018 1 x 5 mL, 200 reactions
4326614 1 x 50 mL 2,000 reactions
4364341 2 x 5 mL, 400 reactions
4364343 5 x 5 mL, 1,000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 4324020 Supplier Applied Biosystems™ Supplier No. 4324020
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

TaqMan™ Universal PCR Master Mix, No AmpErase™ UNG is the ideal reagent solution when you need a Master Mix for multiple 5' nuclease DNA applications.

  • Contains AmpliTaq Gold™ DNA polymerase to provide a better yield and a more robust 5' nuclease assay than AmpliTaq™ DNA polymerase
  • Includes a passive internal reference based on proprietary ROX™ dye, for superb precision on Applied Biosystems™ real-time PCR instruments
  • Buffer enhancements guarantee performance and reliability in all applications, even with G/C-rich sequences
  • Contains dUTP in the dNTP mix to protect against carry-over contamination, when used with AmpErase™ UNG
  • Rapid assay development guidelines are provided to minimize optimization time

Order Info

Guaranteed minimum shelf life is 60 days. Shipping Conditions: Wet Ice.

Specifications

Content And Storage Supplied at 2X concentration. The mix contains AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1 and optimized buffer components. This pack contains 10 X 5 mL tubes, sufficient for 2000 reactions at the 50 μL total volume. Store at 2-8°C

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
Detection Method Primer-probe
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, GeneAmp 9600, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
Passive Reference Dye ROX (Pre-mixed)
Product Line TaqMan
Product Type Universal PCR Master Mix
Quantity 10 x 5 mL, 2,000 reactions
Shipping Condition Wet Ice
Sufficient For 2000 Reactions
For Use With (Application) Gene Expression, Genotyping, miRNA analysis
No. of Reactions 2000 Reactions
Sample Type DNA (Genomic), cDNA
Volume 50 mL
Show More Show Less
What master mix can I use with the TaqMan SNP Genotyping Assays?

We recommend using either the TaqPath ProAmp Master Mix or the TaqMan Genotyping Master Mix. The TaqMan Genotyping Master Mix has the advantage of proven performance with up to 3 days of pre- and post-PCR stability, allowing you to set up plates ahead of time or read the plates later (see the data here, https://tools.thermofisher.com/content/sfs/brochures/cms_039236.pdf) while the TaqPath ProAmp Master Mix can handle samples that may have inhibitors present.

For Research Use Only. Not for use in diagnostic procedures.