missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Universal PCR Master Mix, no AmpErase™ UNG

TaqMan™ Universal PCR Master Mix, No AmpErase™ UNG is the ideal reagent solution when you need a Master Mix for multiple 5' nuclease DNA applications.

Supplier:  Applied Biosystems™ 4324018

Encompass

 View more versions of this product

Catalog No. 4324018


Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Only null left

Description

Description

  • Contains AmpliTaq Gold™ DNA polymerase to provide a better yield and a more robust 5' nuclease assay than AmpliTaq™ DNA polymerase
  • Includes a passive internal reference based on proprietary ROX™ dye, for superb precision on Applied Biosystems™ real-time PCR instruments
  • Buffer enhancements guarantee performance and reliability in all applications, even with G/C-rich sequences
  • Contains dUTP in the dNTP mix to protect against carry-over contamination, when used with AmpErase™ UNG
  • Rapid assay development guidelines are provided to minimize optimization time

Order Info

Guaranteed minimum shelf life is 60 days. Shipping Conditions: Wet Ice.

Specifications

Chemical Identifiers

Primer-probe
qPCR
Standard
ROX (Pre-mixed)
Universal PCR Master Mix
Gene Expression, Genotyping, miRNA analysis
High
AmpliTaq Gold DNA Polymerase
7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, GeneAmp 9600, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
TaqMan
Wet Ice
DNA (Genomic), cDNA

Specifications

Supplied at 2X concentration. The mix contains AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1 and optimized buffer components. This pack contains one 5 mL tube, sufficient for 200 reactions at the 50 μL total volume. Store at 2-8°C

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
High
AmpliTaq Gold DNA Polymerase
7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, GeneAmp 9600, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
TaqMan
1 x 5 mL, 200 reactions
200 Reactions
200 Reactions
5 mL
Primer-probe
qPCR
Standard
ROX (Pre-mixed)
Universal PCR Master Mix
Wet Ice
Gene Expression, Genotyping, miRNA analysis
DNA (Genomic), cDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.