missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ GTXpress™ Master Mix

Combines AmpliTaq™ Fast DNA Polymerase, UP, dNTPs, Tracking Dye, and ROX™ dye in a convenient single vial format.

Supplier:  Applied Biosystems™ 4401892

Encompass

 View more versions of this product

Catalog No. 4401892

Only null left

Includes: Contains 2 x 50mL TaqMan™ GTXpress™ Master Mix (2X master mix containing AmpliTaq™ Fast DNA Polymerase, UP, dNTPs, Tracking Dye and ROX™ Dye).

Description

Description

TaqMan™ GTXpress™ Master Mix delivers accurate genotyping results with robust performance in less than 50 min. The 100mL pack (2 x 50mL) includes reagents sufficient to perform 40,000 PCR reactions (5μL/reaction).

  • Compatible with both Fast and Standard mode thermal cycling conditions
  • Pre-mixed components stored at 2° to 8°C significantly reduce assay setup time
  • Validated with all types of Applied Biosystems™ TaqMan™ Genotyping Assays
  • Extended PCR cycling can be used for poorly amplified or sample-limited reactions The TaqMan™ GTXpress™ Master Mix is also sold as part of TaqMan™ Sample-to-SNP™ Kit, which provides real-time PCR-ready DNA in 5 min. from a wide variety of samples, ranging from blood to buccal swabs to plant tissues. For more information on the TaqMan™ Sample-to-SNP™ Kit, refer to the Applied Biosystems™ TaqMan™ Sample-to-SNP™ Kit Protocol.

Order Info

Guaranteed minimum shelf life is 60 days.

Specifications

Chemical Identifiers

2X
High
qPCR
Genotyping

Specifications

2X
Primer-probe
High
AmpliTaq DNA Polymerase
SNP Genotyping (Real-Time PCR-Based)
SNPs (Unknown or Numerous), SNPs (Known)
GTXpress, TaqMan
UP (Ultra Pure)
Wet Ice
2 X
40,000 Reactions
100 mL
Contains 1 x 10 mL TaqMan™ GTXpress™ Master Mix (2X mix of AmpliTaq™ Fast DNA Polymerase, UP, dNTPs, Tracking Dye and ROX™ Dye). Sufficient for 4,000 reactions. Store at 2-8°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
Single Vial
qPCR
Fast or Standard
QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™ Standard Mode, StepOne™ Fast Mode
ROX (Pre-mixed)
Master Mix
10 mL
Genotyping
Built-In Hot Start
DNA (Genomic), Sample-to-SNP™ Kit Lysates
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.