missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Gene Expression Master Mix

Catalog No. 4370048
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 x 5 mL
2 x 5 mL
5 x 5 mL
10 x 5 mL
1 x 50 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4370048 1 x 1 mL
4369542 10 x 5 mL
4369514 2 x 5 mL
4369510 5 x 5 mL
4369016 1 x 5 mL
4370074 1 x 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 4370048 Supplier Applied Biosystems™ Supplier No. 4370048
Only null left

Requires

Template and primers

Precise and reliable real-time qPCR quantification over 9 logs of linear dynamic range with Applied Biosystems™ TaqMan™ Gene Expression Master Mix.

TaqMan™ Gene Expression Master Mix has been validated with TaqMan™ Gene Expression Assays and Applied Biosystems™ real-time systems, providing precise quantification for a variety of gene expression-based real-time qPCR applications, including rare transcript detection, gene expression analysis, pathogen detection and viral load quantification,co-amplification of two targets, as well as specific detection of gene family members

TaqMan™ Gene Expression Master Mix contains AmpliTaq Gold™ DNA Polymerase, UP (Ultra Pure) for hot start activation and improved detection of bacterial targets, a blend of dNTPs with dTTP/dUTP and Uracil-DNA Glycosylase (UDG) to minimize carry-over PCR contamination, and a passive internal reference based on proprietary ROX™ dye for superb precision on Applied Biosystems™ real-time PCR instruments.

  • Provides precise and reliable real-time qPCR quantification over nine logs of linear dynamic range
  • TaqMan Gene Expression Master Mix is an optimized 2X mix that contains all of the components, excluding the template and primers, for sensitive detection down to one copy of target
  • Extended benchtop stability provides consistent results for automated liquid-handling systems
  • This makes it the choice for high-throughput sample processing overnight or over the weekend
  • Validated with TaqMan gene expression assays and Applied Biosystems real-time systems, providing precise quantification for a variety of gene expression–based real-time qPCR applications
  • Applications include rare transcript detection, gene expression analysis, pathogen detection and viral load quantification, co-amplification of two targets, as well as specific detection of gene family members
  • Store at 2°C to 8°C

TaqMan Gene Expression Master Mix contains:

  • AmpliTaq Gold DNA Polymerase, UP (Ultra Pure), for hot start activation and improved detection of bacterial targets
  • A blend of dNTPs with dTTP/dUTP and uracil-DNA glycosylase (UDG) to minimize carry-over PCR contamination
  • A passive internal reference based on proprietary ROX dye for superb precision on Applied Biosystems real-time PCR instruments

Order Info

Shipping Condition: Wet ice

Specifications

Content And Storage 1 tube (1 mL) 2X TaqMan™ Gene Expression Master Mix sufficient for 40 reactions. Store at 2°C to 8°C.


TaqMan™ Gene Expression Master Mix contains AmpliTaq Gold™ DNA Polymerase (Ultra Pure), Uracil-DNA glycosylase, dNTPs (with dUTP), ROX™ Passive Reference, and optimized buffer components.
Detection Method Primer-probe
Form Liquid
PCR Method qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7500 System, 7900HT System, Applied Biosystems StepOnePlus™ Fast Real-Time PCR SystemStepOne™, Standard Mode, StepOnePlus™, Standard Mode
Passive Reference Dye ROX (Pre-mixed)
Product Line TaqMan
Purity or Quality Grade UP (Ultra Pure)
Quantity 1 x 1 mL
For Use With (Application) Gene Expression
No. of Reactions 40 Reactions
Sample Type DNA (Genomic), cDNA
Volume 1 x 1 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.