missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ SYBR™ Select Master Mix
Cost-effective solution for real-time PCR applications.
$176.00 - $6,620.00
Specifications
| Concentration | 2X |
|---|---|
| Content And Storage | The 2x mix contains SYBR™ GreenER™ dye, AmpliTaq™ DNA Polymerase UP, dNTPs with dUTP/dTTP blend, heat-labile UDG, ROX passive reference dye and optimized buffer components. Contains 1 X 1 mL tube, sufficient for 100, 20 μL reactions (10 μL Master Mix per reaction). Store at 2–8°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). See user guide for primer concentration recommendations for the StepOne and StepOnePlus systems. |
| Detection Method | SYBR |
| Form | Liquid |
| Format | Vial |
Includes: 2X mix contains SYBR GreenER dye, AmpliTaq DNA Polymerase UP, dNTPs with dUTP/dTTP blend, heat-labile UDG, ROX passive reference dye and optimized buffer components.
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
4472903
|
Applied Biosystems™
4472903 |
1 x 1 mL |
Each for $176.00
|
|
|||||
4472908
|
Applied Biosystems™
4472908 |
1 x 5 mL |
Each for $762.00
|
|
|||||
4472918
|
Applied Biosystems™
4472918 |
2 x 5 mL |
Each for $1,456.00
|
|
|||||
4472919
|
Applied Biosystems™
4472919 |
5 x 5 mL |
Each for $3,405.00
|
|
|||||
4472920
|
Applied Biosystems™
4472920 |
10 x 5 mL |
Each for $6,210.00
|
|
|||||
4472913
|
Applied Biosystems™
4472913 |
1 x 50 mL |
Each for $6,620.00
|
|
|||||
Description
- Specific - minimize primer-dimer and nonspecific amplification
- Reproducible and sensitive - consistent amplification across a wide dynamic range
- Bright - contains SYBR™ GreenER™ dye for maximum brightness
- Carry-over contamination control - contains heat-labile UDG
- Can be used in either standard or fast cycling mode
- SYBR™ Select Master Mix contains all the components needed for your real-time PCR reaction, except the template and primers, in a convenient 2X concentration premix
- The master mix includes AmpliTaq™ DNA Polymerase, UP, a highly purified DNA polymerase with a proprietary hot start mechanism that provides exceptional specificity
- SYBR™ Select Master Mix is specially formulated to provide robust results from 100ng to 0.1pg cDNA per reaction and can reliably detect a single copy of genomic DNA
- SYBR™ GreenER™ dye is less inhibitory to PCR than SYBR™ Green I dye resulting in brighter signals
- Heat-labile uracil-DNA glycosylase (UDG) is included for worry-free carryover contamination control
- SYBR™ Select Master Mix can be used in either standard or fast cycling mode and is compatible with all Applied Biosystems™ real-time PCR instruments except the 7900HT real-time PCR system
- It is also compatible with the Bio-Rad IQ™5, Roche LightCycler™ LC480, and Stratagene™ MX3005P™ systems
Order Info
Shipping Conditions: Wet Ice
Specifications
| 2X | |
| SYBR | |
| Vial | |
| qPCR | |
| Fast or Standard | |
| ROX (Pre-mixed) | |
| Real Time PCR SYBR Master Mix | |
| 100 Reactions of 20 μL | |
| Built-In Hot Start |
| The 2x mix contains SYBR™ GreenER™ dye, AmpliTaq™ DNA Polymerase UP, dNTPs with dUTP/dTTP blend, heat-labile UDG, ROX passive reference dye and optimized buffer components. Contains 1 X 1 mL tube, sufficient for 100, 20 μL reactions (10 μL Master Mix per reaction). Store at 2–8°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). See user guide for primer concentration recommendations for the StepOne and StepOnePlus systems. |
|
| Liquid | |
| High | |
| AmpliTaq DNA Polymerase | |
| SYBR GreenER | |
| SYBR | |
| UP (Ultra Pure) | |
| Gene Expression | |
| DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.