missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Fast SYBR™ Green Master Mix

Catalog No. 4385618
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 x 5 mL
2 x 5 mL
5 x 5 mL
10 x 5 mL
1 x 50 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4385618 10 x 5 mL
4385610 1 x 1 mL
4385612 1 x 5 mL
4385616 2 x 5 mL
4385617 5 x 5 mL
4385614 1 x 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 4385618 Supplier Applied Biosystems™ Supplier No. 4385618
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

SYBR Green I Dye, AmpliTaq Gold Fast DNA Polymerase LD, dNTPs with dUTP/dTTP blend, Passive Reference I and optimized buffer components

Requires

Template and primers

Designed for speed, SYBR™ Green Master Mix provides a fast, reliable, and cost-effective solution for RT-PCR application without compromising sensitivity, specificity, dynamic range, or PCR efficiency.

  • A fast, reliable, and cost-effective solution for real-time PCR applications without compromising sensitivity, specificity, dynamic range, or PCR efficiency
  • Real-time PCR results in as fast as 35 minutes
  • Capable of detecting very low copies of target
  • Minimizes primer-dimer and non-specific amplification
  • Consistent amplification across a wide dynamic range
  • Contains all of the components, excluding the template and primers, in a convenient 2X master mix
  • Store at -5°C to -30°C

Includes the following components in an optimized buffer:

  • AmpliTaq™ Fast DNA Polymerase, UP, a highly purified DNA polymerase designed to allow instant hot start
    • Minimizes non-specific product formation and enables reactions to be set up at room temperature
  • SYBR™ Green I dye to enable detection of double-stranded DNA
  • Deoxynucleotides (dNTPs) to help maintain optimal PCR results
  • Uracil-DNA Glycosylase (UDG) designed to reduce carryover contamination
  • Passive internal reference based on proprietary ROX™ dye to enable increased precision

Order Info

Shipping Condition: Dry Ice

Specifications

Concentration 2X
Content And Storage 10 × 5 mL vials. The 2X mix contains SYBR™ Green 1 Dye, AmpliTaq Gold™ Fast DNA Polymerase LD, dNTPs with dUTP⁄dTTP blend, Passive Reference 1, and optimized buffer components. Sufficient for 5000 reactions of 20 μL each.

Store at -5 to -30°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Detection Method SYBR
Format Vial
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase AmpliTaq Fast DNA Polymerase, AmpliTaq Fast UP
Reaction Speed Fast
For Use With (Equipment) 7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
Label or Dye SYBR Green
Passive Reference Dye ROX (Pre-mixed)
Product Line SYBR
Product Type Real Time PCR SYBR Master Mix
Purity or Quality Grade UP (Ultra Pure)
Quantity 10 x 5 mL
Sufficient For 5000 Reactions of 20 μL Each
For Use With (Application) Gene Expression
No. of Reactions 5000 Reactions
Sample Type DNA (Genomic), cDNA
Volume 10 x 5 mL
Show More Show Less
Is there a SYBR Green Fast Master Mix currently available?

Yes. The following reagents can be used for fast cycling: Fast SYBR Green Master Mix, EXPRESS SYBR GreenER qPCR Supermix Universal, and SYBR GreenER qPCR SuperMix for ABI PRISM instruments.

For Research Use Only. Not for use in diagnostic procedures.