missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Fast SYBR™ Green Master Mix

Designed for speed, SYBR™ Green Master Mix provides a fast, reliable, and cost-effective solution for RT-PCR application without compromising sensitivity, specificity, dynamic range, or PCR efficiency.

Supplier:  Applied Biosystems™ 4385612

Encompass

 View more versions of this product

Catalog No. 4385612


Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Only null left

Includes: SYBR Green I Dye, AmpliTaq Gold Fast DNA Polymerase LD, dNTPs with dUTP/dTTP blend, Passive Reference I and optimized buffer components

Requires : Template and primers

Description

Description

  • A fast, reliable, and cost-effective solution for real-time PCR applications without compromising sensitivity, specificity, dynamic range, or PCR efficiency
  • Real-time PCR results in as fast as 35 minutes
  • Capable of detecting very low copies of target
  • Minimizes primer-dimer and non-specific amplification
  • Consistent amplification across a wide dynamic range
  • Contains all of the components, excluding the template and primers, in a convenient 2X master mix
  • Store at -5°C to -30°C

Includes the following components in an optimized buffer:

  • AmpliTaq™ Fast DNA Polymerase, UP, a highly purified DNA polymerase designed to allow instant hot start
    • Minimizes non-specific product formation and enables reactions to be set up at room temperature
  • SYBR™ Green I dye to enable detection of double-stranded DNA
  • Deoxynucleotides (dNTPs) to help maintain optimal PCR results
  • Uracil-DNA Glycosylase (UDG) designed to reduce carryover contamination
  • Passive internal reference based on proprietary ROX™ dye to enable increased precision

Order Info

Shipping Condition: Dry Ice

Specifications

Chemical Identifiers

2X
High
AmpliTaq Fast DNA Polymerase, AmpliTaq Fast UP
7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
ROX (Pre-mixed)
Real Time PCR SYBR Master Mix
Gene Expression
SYBR
qPCR
Fast
SYBR Green
SYBR
UP (Ultra Pure)
DNA (Genomic), cDNA

Specifications

2X
SYBR
High
AmpliTaq Fast DNA Polymerase, AmpliTaq Fast UP
7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
ROX (Pre-mixed)
Real Time PCR SYBR Master Mix
1 x 5 mL
500 Reactions of 20 μL Each
500 Reactions
5 mL
Contains 1 × 5 mL vial. The 2X mix contains SYBR™ Green 1 Dye, AmpliTaq Gold™ Fast DNA Polymerase LD, dNTPs with dUTP/dTTP blend, Passive Reference 1, and optimized buffer components. Sufficient for 500 reactions of 20 μL each.

Store at -5 to -30°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Vial
qPCR
Fast
SYBR Green
SYBR
UP (Ultra Pure)
Dry Ice
Gene Expression
DNA (Genomic), cDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.