missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ Optical Adhesive Film

Reduces chance of well-to-well contamination and sample evaporation when applied to a microplate

$159.00 - $532.00

Specifications

Color Optical
Material Polyester
For Use With (Equipment) 2720 Thermal Cycler, 6100 Thermal Cycler, 7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp 9700, QuantStudio™, SimpliAmp™ Thermal Cycler, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System, ViiA™ 7 Dx Fast System, QuantStudio™ Dx, 7500 Fast Dx System, ViiA™ 7 Fast System, ProFlex™ PCR System
Shipping Condition Room Temperature
Product Type Film, Seal, or Applicator
View More Specs

Includes: 100 Optical Adhesive Covers

Products 2
Catalog Number Mfr. No. No. per Case Price Quantity & Availability  
Catalog Number Mfr. No. No. per Case Price Quantity & Availability  
4311971
Encompass
View Documents Promotion Details
Applied Biosystems™
4311971
100 Covers
Each for $532.00
Only null left
 
4360954
Encompass
View Documents Promotion Details
Applied Biosystems™
4360954
25 Covers
Each for $159.00
Only null left
 
Description

Description

Applied Biosystems MicroAmp Optical Adhesive Film is designed to reduce well-to-well contamination and sample evaporation. The easy-to-use film is applied by peeling away the backing and firmly pressing the adhesive film across the top of a microplate. This tight seal will help ensure consistent real-time PCR data without obscuring samples during reading, and the protective covering allows microplates to be more easily stacked for space-saving convenience and storage.

Features

  • Pressure-sensitive adhesive provides a tight seal over every well
  • Will not interfere with sample reading
  • Easy-to-apply film will not stick to gloves
  • Prevents sample loss and reduces cross-contamination
  • Designed to fit MicroAmp 96- and 384-well plates

Notes

Thermo Fisher Scientific has performed exhaustive functional testing regarding minor visual inconsistencies (e.g., wrinkles, line impressions, haziness, white spots, etc.) of the MicroAmp Optical Adhesive Film that may result from removal of the liner or application to a plate. These minor visual inconsistencies do not affect the performance of the adhesive tape. Should you have any additional questions, please contact Technical Support.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Optical
2720 Thermal Cycler, 6100 Thermal Cycler, 7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp 9700, QuantStudio™, SimpliAmp™ Thermal Cycler, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System, ViiA™ 7 Dx Fast System, QuantStudio™ Dx, 7500 Fast Dx System, ViiA™ 7 Fast System, ProFlex™ PCR System
Film, Seal, or Applicator
Pressure Sensitive
Polyester
Room Temperature
MicroAmp
96/384-well Plate
Videos
SDS
Documents

Documents