missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ Optical Adhesive Film

Catalog No. 4360954
Encompass
Change view
Click to view available options
No. per Case:
100 Covers
25 Covers
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Case
4360954 25 Covers
4311971 100 Covers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4360954 Supplier Applied Biosystems™ Supplier No. 4360954

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

100 Optical Adhesive Covers

Reduces chance of well-to-well contamination and sample evaporation when applied to a microplate

Applied Biosystems MicroAmp Optical Adhesive Film is designed to reduce well-to-well contamination and sample evaporation. The easy-to-use film is applied by peeling away the backing and firmly pressing the adhesive film across the top of a microplate. This tight seal will help ensure consistent real-time PCR data without obscuring samples during reading, and the protective covering allows microplates to be more easily stacked for space-saving convenience and storage.

Features

  • Pressure-sensitive adhesive provides a tight seal over every well
  • Will not interfere with sample reading
  • Easy-to-apply film will not stick to gloves
  • Prevents sample loss and reduces cross-contamination
  • Designed to fit MicroAmp 96- and 384-well plates

Notes

Thermo Fisher Scientific has performed exhaustive functional testing regarding minor visual inconsistencies (e.g., wrinkles, line impressions, haziness, white spots, etc.) of the MicroAmp Optical Adhesive Film that may result from removal of the liner or application to a plate. These minor visual inconsistencies do not affect the performance of the adhesive tape. Should you have any additional questions, please contact Technical Support.

Order Info

Shipping Condition: Room Temperature

Specifications

Color Optical
Material Polyester
No. per Case 25 Covers
Quantity 25 covers
Content And Storage Includes 25 Optical Adhesive Covers. Store at room temperature.
For Use With (Equipment) 2720 Thermal Cycler, 6100 Thermal Cycler, 7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp 9700, QuantStudio™, SimpliAmp™ Thermal Cycler, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System, ViiA™ 7 Dx Fast System, QuantStudio™ Dx, 7500 Fast Dx System, ViiA™ 7 Fast System, ProFlex™ PCR System
Shipping Condition Room Temperature
Product Type Film, Seal, or Applicator
Product Line MicroAmp
Adhesive Pressure Sensitive
Plate Compatibility 96/384-well Plate
Show More Show Less
How do I correctly apply MicroAmp Optical Adhesive Covers to an optical 96- or 384-well plate?

Please follow the instructions below for the proper application of MicroAmp Optical Adhesive Covers (Cat. Nos. 4311971, 4313663 and 4360954).

The MicroAmp Optical Adhesive Cover uses a pressure-sensitive adhesive backing to adhere the cover to the optical 96- or 384-well plate. It is imperative that you use enough force to activate the pressure-sensitive adhesive to prevent evaporation from the wells.
Please note: Users should not expect the Optical Adhesive Cover to be very sticky to the touch on the side that adheres to the optical plate.

The correct way to apply the cover is as follows:

1. Apply firm pressure when using the Adhesive Seal Applicator (Cat. No. 4333183) to press the cover onto the plate.

2. Run the applicator firmly from left to right several times. Run the applicator up and down several times.

3. Run the edge of the applicator around the border (following the red line outside the wells) of the cover.

4. Repeat the left to right and up and down motions to assure a good final seal.

5. Inspect each well to make sure that each well edges are imprinted on the optical adhesive cover.
Please Note: Using the Applicator's edge to seal between the wells is NOT recommended. It may actually pull the seal off the wells.

Which of your plastic consumables are compatible with the Applied Biosystems 7900HT Fast Real-Time PCR System?

There are many of our reagents and plastic consumables that can be used on the Applied Biosystems 7900HT Fast Real-Time PCR System. Here are a few examples to help you get started:
Cat. No. 4313663, MicroAmp Optical Adhesive Film Kit
Cat. No. 4311971, 4360954, MicroAmp Optical Adhesive Film
Cat. No. 4323032, MicroAmp Optical 8-Cap Strip
Cat. No. 4309849; 4326270, MicroAmp Optical 384-Well Reaction Plate with Barcode
Cat. No. 4306737; 4326659; 4314320, MicroAmp Optical 96-Well Reaction Plate with Barcode
Cat. No. 4346906, MicroAmp Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL
Cat. No. 4312063, Splash Free 96-Well Base

For more comprehensive lists of compatible plastics, enter "Real-Time PCR Plastics" in the main Search field on our website and follow the links.

Can the MicroAmp Optical Adhesive Film be used with MicroAmp Optical Tubes?

No. The Optical Adhesive Film is recommended for use only with the MicroAmp Optical 96-well Reaction Plate (with or without barcode) and MicroAmp 384-well Optical Reaction Plate.