missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Power SYBR™ Green PCR Master Mix

Catalog No. 4368708
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 x 5 mL
2 x 5 mL
5 x 5 mL
10 x 5 mL
1 x 50 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4368708 10 x 5 mL
4368577 1 x 1 mL
4367659 1 x 5 mL
4368706 2 x 5 mL
4368702 5 x 5 mL
4367660 1 x 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 4368708 Supplier Applied Biosystems™ Supplier No. 4368708
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

SYBR™ Green 1 Dye, AmpliTaq Gold DNA Polymerase LD, dNTPs with dUTP/dTTP blend, Passive Reference 1, and optimized buffer components

Requires

Template and primers

Offers superior sensitivity and reproducibility, detecting as few as two copies of a target gene over a broad range of template concentrations.

Get superior nucleic acid quantitation with Applied Biosystems™ Power SYBR™ Green Master Mix.

  • Everything needed for SYBR™ Green dye-based PCR amplification and detection in a convenient single-tube format
  • Power SYBR™ Green PCR Master Mix contains all of the components (excluding the template and primers) for superior SYBR™ Green reagent-based real-time PCR
  • The optimized formulation contains highly purified AmpliTaq Gold™ DNA Polymerase, LD to offer increased sensitivity
  • Contains a blend of dTTP/dUTP that is compatible with AmpErase™ UNG to minimize carryover contamination
  • Includes a proprietary version of ROX™ dye, an internal passive reference, to normalize non-PCR–related fluorescence fluctuations
  • This minimizes well-to-well variability that can result from a variety of causes, such as pipetting error and sample evaporation
  • Power SYBR™ Green reagent-based PCR chemistry easily replaces the existing SYBR™ Green PCR Master Mix in Applied Biosystems™ protocols using the same setup and thermal cycling conditions
  • Store at -20°C

Order Info

Shipping Condition: Wet Ice

Specifications

Concentration 2X
Content And Storage The 2X mix contains SYBR™ Green 1 Dye, AmpliTaq Gold™ DNA Polymerase LD, dNTPs with dUTP/dTTP blend, Passive Reference 1, and optimized buffer components. Contains 10 × 5 mL bottles, sufficient for 2000 reactions of 50 μL each.

Store at -20°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Detection Method SYBR
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase AmpliTaq Gold DNA Polymerase, AmpliTaq Gold LD
Reaction Speed Standard
For Use With (Equipment) 7300 System, 7500 System, 7700 System, 7900HT Fast System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System
Label or Dye SYBR Green
Passive Reference Dye ROX (Pre-mixed)
Product Line SYBR
Product Type Real Time PCR SYBR Master Mix
Purity or Quality Grade LD (Low DNA)
Quantity 10 x 5 mL
Sufficient For 2000 Reactions of 50 μL each
For Use With (Application) Gene Expression
No. of Reactions 2000 Reactions
Sample Type DNA (Genomic), cDNA
Volume 10 x 5 mL
Show More Show Less
What can I do to improve the sensitivity of my qPCR assay?

If you are targeting a low-abundance gene, you may have trouble getting Ct values in a good, reliable range (Ct > 32). To increase the sensitivity of the assay, you may want to consider the following:

- Increase the amount of RNA input into your reverse transcription reaction, if possible
- Increase the amount of cDNA in your qPCR reaction (20% by volume max)
- Try a different reverse transcription kit, such as our SuperScript VILO Master Mix, for the highest cDNA yield possible
- Consider trying a one-step or Cells-to-CT type workflow (depending on your sample type)

How do I set the baseline for my qPCR experiment?

Most times your instrument software can automatically set a proper baseline for your data. Check out our short video, Understanding Baselines, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=5BjFAJHW-bE).

How do I set the threshold for my qPCR experiment?

In most cases your instrument software can automatically set a proper threshold for your data. Check out our short video, Understanding Thresholds, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=H_xsuRQIM9M).

I am not getting any amplification with my TaqMan Assay or SYBR Green primer set. What is causing this?

There could be several reasons for no amplification from an assay or primer set. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/no-amplification.html) for more details.

I am getting amplification in my no-template control (NTC) wells in my qPCR experiment. What is causing this?

There could be several reasons for amplification in a NTC well. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/amplification-no-template-control.html) for more details.

My amplification curves have a funny shape in my qPCR experiment. What is causing this?

There are several reasons that amplification could be delayed. Please see the information in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/abnormal-amplification-curves/amplification-occurs-later.html) for more details.

What can I do if the amplification of my target gene is later than expected for my qPCR experiment?

There are several reasons that amplification could be delayed. Please see the information in our Real-Time Troubleshooting Tool for more details (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/abnormal-amplification-curves/amplification-occurs-later.html).

Can I use my SYBR Green primers for a TaqMan assay?

It may be possible to use your SYBR Green primers for a TaqMan assay, depending on how they were designed. You would have to design a separate probe to use with your existing primers. Please refer to the guidelines in this manual (https://tools.thermofisher.com/content/sfs/manuals/cms_041902.pdf) on “Manually Designing Primers and Probes” for the next steps. If you have Primer Express Software, you can use that software to design a probe. Please note that restricting the design using the predesigned SYBR primers may not allow for a successful probe design.

Do I have to normalize my samples for comparative Ct experiments?

Comparative Ct experiments use an endogenous control gene to normalize the cDNA input. Please watch this short video (https://www.youtube.com/watch?feature=player_embedded&v=jst-3hD_xFQ) for more details on how this works. For a protocol workflow, please refer to our Guide to Performing Relative Quantitation of Gene Expression (https://tools.thermofisher.com/content/sfs/manuals/cms_042380.pdf).

What are the requirements for a relative quantification qPCR experiment?

In a relative quantification experiment, you will need to identify an endogenous control and a reference (or calibrator) sample. An endogenous control is a gene that does not change in expression across all the samples in your study. A reference sample is the sample that you are comparing all others to. This is often the untreated, or control, sample. Please see our Relative Gene Expression Workflow bulletin (https://tools.thermofisher.com/content/sfs/brochures/cms_075428.pdf) for more step-by-step guidelines on how to design your experiment.

What are the requirements for a standard curve qPCR experiment?

In a standard curve experiment, you must generate a standard curve for each target gene. The standards should closely represent the sample (i.e., RNA for RNA input, plasmid or gDNA for DNA input). This reference (http://www.ncbi.nlm.nih.gov/pubmed/11013345) is a good review of standard curves and the experimental setup. You can also review this short video (https://www.youtube.com/watch?v=mE5ieko9_RQ) on standard curve experiments.

What is the difference between absolute quantification (AQ) and relative quantification (RQ)? How do I choose which method to use?

Absolute quantification will quantitate unknowns based on a known quantity. It involves the creation of a standard curve from a target of known quantity (i.e., copy number). Unknowns can then be compared to the standard curve and a value can be extrapolated. Absolute quantification is useful for quantitating copy number of a certain target in DNA or RNA samples. The result usually is a number followed by a unit, such as copy number and ng, etc.

Relative quantification can quantitate a fold difference between samples. It involves the comparison of one sample to another sample (calibrator) of significance. For example, in a drug treatment study you could compare a treated to an untreated sample. The quantity of the calibrator is not known and cannot be measured absolutely. Therefore the calibrator (untreated sample) and samples (treated samples) are normalized to an endogenous control (a gene that is consistently expressed among the samples) and then compared to each other to get a fold difference. Relative quantification is useful for quantitating messenger RNA levels. Since the result is a fold change or ratio, it is not followed by a unit.

The method that you choose will depend on the type of data you need from your experiment. You can find more information here (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/absolute-vs-relative-quantification-for-qpcr.html) as well.

Can I do a melt curve with a TaqMan assay?

No. A TaqMan probe, once cleaved, cannot be re-quenched. Therefore a melt curve does not apply when using a TaqMan assay.

What is the difference between TaqMan and SYBR Green methods of detection?

TaqMan and SYBR Green chemistries are two different methods of detection for qPCR. Please see this detailed comparison of these two approaches (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/taqman-assays-vs-sybr-green-dye-for-qpcr.html). You can also watch this short video (https://www.youtube.com/watch?feature=player_embedded&v=fkUDu042xic) on how TaqMan assays work.

How many replicates do I need to run for my qPCR experiment? What recommendations do you have for plate setup?

Please view this short video (https://www.youtube.com/watch?v=eIaPGhOjBQo), which explains some best practices for replicates and plate setup.

What are the different phases of a qPCR reaction?

Check out this short video (https://www.youtube.com/watch?feature=player_embedded&v=4sXPUbIrh3A) to understand the different phases of the PCR reaction and why they are important.

I'm trying to decide between purchasing a one-step or two-step RT-PCR kit. Can you review the advantages and disadvantages of each?

One-step RT-PCR is convenient and less prone to contamination, as there is less opportunity for pipetting error. This method is also faster than the two-step process. However, the cDNA cannot be archived, and fewer genes can be analyzed. Two-step RT-PCR gives you the ability to archive cDNA, analyze multiple genes, and offers greater flexibility. Learn more about the difference between one-step and two-step RT-PCR on this page Onestep vs Twostep RT-PCR.

Can Power SYBR Green PCR Master Mix be substituted directly for the SYBR Green PCR Master Mix?

The Power SYBR Green PCR Master Mix (P/N 4368577/ 4367659/ 4368706/ 4368702/ 4368708/ 4367660) is a new and improved formulation of the regular SYBR Green PCR Master Mix (P/N 4344463/ 4309155/ 4364344/ 4364346/ 4312704/ 4334973). This new master mix contains a more purified AmpliTaq enzyme and improved buffer formulation compared to regular SYBR Green PCR Master Mix. You may need to re-validate your assays in order to start using the Power SYBR Green PCR Master Mix. For more information on validating your reaction with the Power SYBR Green PCR Master Mix, please refer to the Power SYBR Green PCR Master Mix Protocol (P/N 4367218).

Can Power SYBR Green PCR Master Mix (Cat. No. 4368577, 4367659, 4368706, 4368702, 4368708, 4367660) be substituted directly for the SYBR Green PCR Master Mix (Cat. No. 4309155, 4364344, 4364346, 4312704, 4334973)?

The Power SYBR Green PCR Master Mix is a new and improved formulation of the regular SYBR Green PCR Master Mix. You may need to verify your assays in order to start using the Power SYBR Green PCR Master Mix. For more information on verifying your reaction with the Power SYBR Green PCR Master Mix, please refer to the Power SYBR Green PCR Master Mix protocol.

How can I reduce nonspecific amplification with the Power SYBR green Master Mix (Cat. No. 4368577, 4367659, 4368706, 4368702, 4368708, 4367660)?

For one-step RT-PCR, this protocol requires an initial incubation of the reaction mixture for 30 minutes at 48°C. This RT step incubates the PCR primers at a temperature below their annealing temperatures. The AmpliTaq Gold DNA Polymerase enzyme will slowly activate at 48°C and may lead to non-specific amplification. To minimize the level of nonspecific amplification in One-Step RT-PCR using Power SYBR Green PCR Master Mix, lower primer concentrations are recommended. If non-specific amplification is still problematic, reverting to Two-Step RT-PCR is recommended. For optimizing primer concentrations see Plate configuration for Primer Optimization for One-step RT-PCR.

Does Power SYBR Green PCR Master Mix (Cat. No. 4368577, 4367659, 4368706, 4368702, 4368708, 4367660) work with reverse transcription (RT)-PCR?

Yes, the Power SYBR Green RT-PCR Reagents Kit contains all of the components for either one-step or two-step RT-PCR. For one-step RT-PCR, reverse transcriptase and RNase inhibitor are added to Power SYBR Green PCR Master Mix before cycling begins. For two-step RT-PCR, RNA is first reverse transcribed into cDNA and then the resulting cDNA is quantitated in the second step using the Power SYBR Green PCR Master Mix.

I received the Power SYBR Green Master Mix (Cat. No. 4368577, 4367659, 4368706, 4368702, 4368708, 4367660) and it appeared to have an orange color. Is this normal?

It is normal for the Power SYBR Green Master Mix to have an orange hue. It is due to the SYBR green dye.

How can I use the Power SYBR Green Master Mix (Cat. No. 4368577, 4367659, 4368706, 4368702, 4368708, 4367660) in Fast mode?

In Fast mode with the Power SYBR Green Master Mix, use a 20 µL total reaction volume per well on the 7500 Fast or 7900HT systems. The cycling conditions are as follows:

  • Hold: 95°C/10 min
  • Denaturation: 95°C/3 sec
  • Anneal/Extension: 60°C/30 sec

Can the Power SYBR Green Master Mix (Cat. No. 4368577, 4367659, 4368706, 4368702, 4368708, 4367660) be used in Fast mode? Which instruments have been tested in Fast mode?

Yes. We have tested the Power SYBR Green Master Mix in the Fast mode on the 7500 Fast and 7900HT real time PCR instruments.

Why does Power SYBR Green have different storage conditions compared to existing SYBR?

Power SYBR Green has excellent stability. However, due to the nature of SYBR Green dye (sensitivity to light and temperature shift), the recommended storage condition of Power SYBR Green Master Mix is -15°C-20°C for long-term storage. You can store Power SYBR Green at 2-8°C up to 6 months.

What is the shelf life of Power SYBR Green PCR Master Mix (Cat. No. 4368577)?

Power SYBR Green PCR Master Mix has a guaranteed minimum shelf life of 60 days. The exact expiry date can be found in the CofA.

What is the difference in sensitivity between TaqMan chemistry vs. SYBR Green reagent chemistry?

Sensitivity can actually be equivalent when using TaqMan chemistry and SYBR Green reagent chemistry. It might seem that a TaqMan assay with fluorescent signal generated by a sequence-specific probe would always be more sensitive than a SYBR Green reagent assay, but a poorly designed TaqMan assay could theoretically be less specific than a well-designed SYBR Green reagent assay. However, the potential for detection of primer dimers and non-specific products using SYBR Green chemistry is more likely to result in loss of sensitivity when attempting to quantitate lower copy numbers.

For more information on Real-Time PCR chemistries, please refer to the following Application Notes, which you can find on our website through Technical Resources, or by entering the titles in the main Search field: “Real-Time PCR Vs. Traditional PCR”, “Essentials of Real Time PCR”, and “Selection of Reagents for Real-Time PCR”.

What are the key differences between a TaqMan MGB probe and a TaqMan TAMRA dye-labeled probe?

The TaqMan MGB probes contain the following features:
1) A fluorescent reporter at the 5' end
2) A nonfluorescent quencher at the 3' end. Because the quencher does not fluoresce, the real-time instruments can measure the reporter dye contributions more precisely.
3) A minor groove binder at the 3' end. The minor groove binder increases the melting temperature (Tm) of probes, allowing the use of shorter probes.

In general, the TaqMan MGB probes exhibit great differences in Tm values between matched and mismatched probes, which provides more accurate allelic discrimination and makes for a more sensitive real-time assay. Mismatches between a probe and allele, or target, reduce the efficiency of probe hybridization in a measurable way, which is especially important in SNP Genotyping assays. Furthermore, AmpliTaq Gold DNA polymerase is more likely to displace the mismatched probe rather than cleave it to release reporter dye. More information about TaqMan MGB probes can be found in the User Bulletin entitled "Primer Express Version 1.5 and TaqMan MGB Probes for Allelic Discrimination." You can find a copy on our website by entering this title in the main search field.

When using SYBR Green chemistry on an Applied Biosystems Real-Time PCR instrument, how do I change settings to reflect that there is no TaqMan probe being used in the reaction?

Refer to the product manual for your instrument and software for specifics, but in general you will want to change the Quencher value to None.

How can RNA standards be generated to perform absolute quantitation for RNA targets?

It is generally not possible to use DNA as a standard for absolute quantitation of RNA because there is no control for the efficiency of the reverse transcription step. Therefore, in-vitro transcribed RNA is commonly used to prepare standards for the absolute quantitation of RNA targets. This would involve the cloning of the target of interest into an in-vitro transcription plasmid, performing in-vitro transcription, then purifying the resulting cRNA so that the DNA plasmid cannot serve as a PCR template. Concentration is measured by A260 and converted to the number of copies using the molecular weight of the RNA.

Relative quantitation of gene expression methods require less up-front preparation and provide a fold-change value instead of an absolute quantity result. For many researchers, absolute quantities are not a necessary parameter to measure, and therefore relative quantitation is a much more attractive approach to studying gene expression via real-time PCR. For more information on relative quantitation of gene expression, please refer to our Technical Reference Library in the Technical Resources section of our website.


For Research Use Only. Not for use in diagnostic procedures.