Learn More
Applied Biosystems™ AmpliTaq™ DNA Polymerase with Buffer I or Buffer II
AmpliTaq DNA Polymerase is a 94 kDa, thermostable, recombinant DNA polymerase obtained by expression of a modified form of the Thermus aquaticus (Taq) DNA polymerase gene in E. coli.
$340.00 - $18,310.00
Specifications
| Concentration | 5 U/μL |
|---|---|
| Format | Stand-alone enzyme |
| Polymerase | AmpliTaq DNA Polymerase |
| Reaction Speed | Standard |
| Exonuclease Activity | 5' - 3' |
Includes: Contains 1000 units of AmpliTaq DNA Polymerase, Buffer II, and MgCl2 Solution.
| Catalog Number | Mfr. No. | Includes | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Quantity | Price | Quantity & Availability | ||||
N8080161
|
Applied Biosystems™
N8080161 |
Buffer II | 250 Units |
Each for $340.00
|
|
||||
N8080172
|
Applied Biosystems™
N8080172 |
Buffer II | 1000 Units |
Each for $1,326.00
|
|
||||
N8080153
|
Applied Biosystems™
N8080153 |
Buffer II | 3,000 units |
Each for $3,355.00
|
|
||||
N8080156
|
Applied Biosystems™
N8080156 |
Buffer II | 5,000 units |
Each for $4,885.00
|
|
||||
N8080186
|
Applied Biosystems™
N8080186 |
Buffer II | 25,000 units |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
N8080152
|
Applied Biosystems™
N8080152 |
Buffer I | 3,000 units |
Each for $3,335.00
|
|
||||
N8080155
|
Applied Biosystems™
N8080155 |
Buffer I | 5,000 units |
Each for $4,085.00
|
|
||||
N8080160
|
Applied Biosystems™
N8080160 |
Buffer I | 250 Units |
Each for $346.00
|
|
||||
N8080171
|
Applied Biosystems™
N8080171 |
Buffer I | 1,000 units |
Each for $1,336.00
|
|
||||
N8080185
|
Applied Biosystems™
N8080185 |
Buffer I | 25,000 Units |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
AmpliTaq DNA Polymerase is a 94-kDa, thermostable, recombinant DNA polymerase obtained by expression of a modified form of the Thermus aquaticus (Taq) DNA polymerase gene in E. coli. It is the most thoroughly characterized enzyme available for the PCR process, and its recombinant nature and purification method provide unparalleled purity and reproducibility, vial-to-vial and lot-to-lot.
AmpliTaq DNA Polymerase is available with GeneAmp10X PCR Buffer I or with GeneAmp 10X PCR Buffer II and MgCl2 Solution.
Reliable and robust PCR
The thermal activity profile of AmpliTaq DNA Polymerase is good for PCR applications because its optimal activity is in the same range at which stringent annealing of primers occurs (55–75°C). The enzyme's half-life is ∼40 minutes at 95°C, providing thermostability that meets the requirements of most difficult PCR applications.
Features
- The most thoroughly characterized enzyme available for the PCR process
- Excellent thermal activity profile makes it good for PCR applications
- QC-tested to guarantee reproducible results
Notes
- For superior PCR performance, we recommend DreamTaq DNA Polymerase.
Order Info
Shipping Condition: Dry ice
Specifications
| 5 U/μL | |
| AmpliTaq DNA Polymerase | |
| 5' - 3' | |
| Dry Ice | |
| 1X | |
| 3'-A | |
| 5 kb or less |
| Stand-alone enzyme | |
| Standard | |
| DNA Polymerase | |
| Standard PCR | |
| No | |
| Separate Components | |
| DNA |
For Research Use Only. Not for use in diagnostic procedures.