missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ apHera™ NH2 HPLC Guard Column, 5 μm Particle Size

Catalog No. 111016065
Change view
Click to view available options
D: 2 mm; L: 1 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111016065 D: 2 mm; L: 1 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016065 Supplier MilliporeSigma™ Supelco™ Supplier No. 56429AST

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with apHera™ HPLC columns

Specifications

Quantity 1/Pk.
Particle Size 5 μm
Length (Metric) 1 cm
Pore Size 300 Å
Type Guard Column
Diameter (Metric) Inner 2 mm
For Use With (Application) HPLC
Column Type HPLC Guard Column
Product Line apHera

apHera is a trademark of Sigma-Aldrich Co. LLC