missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Anza™ DNA Blunt End Kit

Catalog No. IVGN2404
Encompass
Change view
Click to view available options
Quantity:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IVGN2404 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IVGN2404 Supplier Invitrogen™ Supplier No. IVGN2404

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Anza™ Blunting Enzyme Mix, Anza™ 10X Blunting Buffer,

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
  • Part of the Anza™ Restriction Enzyme Cloning System
  • Use directly following heat inactivation of digestion reaction
  • Quick 15 minute protocol

Certifications

CoA

Specifications

Content And Storage 100 μL Anza Blunting Enzyme Mix
200 μL Anza 10X Blunting Buffer

Store at -5 to -30°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Enzyme T4 DNA polymerase, Klenow Fragment
Compatible Buffer Anza™ 10X Blunting Buffer
Quantity 100 reactions
Product Type DNA Blunt End Kit
Product Line Anza

For Research Use Only. Not for use in diagnostic procedures.