missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Antibodix™ WCX-NP5 Weak Cation Exchange Guard Cartridge and Holder, 5 μm Particle Size, Stainless Steel

Catalog No. 111008524
Change view
Click to view available options
Diameter (Metric) Inner:
2 mm
4 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
111008524 4 mm
111008523 2 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111008524 Supplier MilliporeSigma™ Supelco™ Supplier No. Z777291

Antibodix™ columns are designed for the reproducible analysis of charge variants of monoclonal antibodies by salt or pH gradient.

The non-porous poly(styrene divinylbenzene) particles in Antibodix™ columns are shielded by a hydrophilic polymer layer that contains a network of weak (WCX) cation exchange groups.

Specifications

pH 2.0 to 12.0
Particle Size 5 μm
Length (Metric) 1 cm
Pore Size Non-porous
Type Cation Exchange HPLC Guard Cartridge and Holder
Diameter (Metric) Inner 4 mm
Material Stainless Steel
Phase Cation Exchange
For Use With (Application) HPLC
Product Line Antibodix

Antibodix is a registered trademark of Sepax Technologies