missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Antibodix™ WCX-NP3 Weak Cation Exchange Column, PEEK, 3 μm Particle Size

Catalog No. 111006438
Change view
Click to view available options
Diameter (Metric):
2.1 mm
4.6 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
111006438 2.1 mm
111006439 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111006438 Supplier MilliporeSigma™ Supelco™ Supplier No. Z777287
Only null left

Antibodix™ columns are designed for the reproducible analysis of charge variants of monoclonal antibodies by salt or pH gradient.

The non-porous poly(styrene divinylbenzene) particles in Antibodix™ columns are shielded by a hydrophilic polymer layer that contains a network of weak (WCX) cation exchange groups.

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 5 cm
Particle Size 3 μm
pH 2.0 to 12.0
Pore Size Nonporous
Stationary Phase Spherical PS-DVB
Product Line Antibodix
Matrix Spherical PS-DVB Phase
Phase Cation Exchange
Material PEEK
Technique Cation Exchange
Show More Show Less

Antibodix is a registered trademark of Sepax Technologies