missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Antibodix™ WCX-NP10 Weak Cation Exchange Column, 10 μm Particle Size, Stainless Steel

Catalog No. 111008220
Change view
Click to view available options
Length (Metric):
5 cm
25 cm
Diameter (Metric):
2.1 mm
4.6 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
111008220 5 cm 2.1 mm
111006369 25 cm 2.1 mm
111008221 5 cm 4.6 mm
111006451 25 cm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111008220 Supplier MilliporeSigma™ Supelco™ Supplier No. Z777268

Antibodix™ columns are designed for the reproducible analysis of charge variants of monoclonal antibodies by salt or pH gradient.

The non-porous poly(styrene divinylbenzene) particles in Antibodix™ columns are shielded by a hydrophilic polymer layer that contains a network of weak (WCX) cation exchange groups.

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 5 cm
Particle Size 10 μm
pH 2.0 to 12.0
Pore Size Nonporous
Stationary Phase Spherical PS-DVB
Product Line Antibodix
Matrix Spherical PS-DVB Phase
Phase Cation Exchange
Material Stainless Steel
Technique Cation Exchange
Show More Show Less

Antibodix is a registered trademark of Sepax Technologies