missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Antibodix™ WCX-NP1.7 Weak Cation Exchange Guard Cartridge and Holder, 1.7 μm Particle Size, Stainless Steel
Antibodix™ columns are designed for the reproducible analysis of charge variants of monoclonal antibodies by salt or pH gradient.
$1293.26 - $1293.26
Specifications
| pH | 2.0 to 12.0 |
|---|---|
| Particle Size | 1.7 μm |
| Length (Metric) | 1 cm |
| Pore Size | Non-porous |
| Type | Cation Exchange Guard Cartridge and Holder |
Description
The non-porous poly(styrene divinylbenzene) particles in Antibodix™ columns are shielded by a hydrophilic polymer layer that contains a network of weak (WCX) cation exchange groups.Specifications
| 2.0 to 12.0 | |
| 1 cm | |
| Cation Exchange Guard Cartridge and Holder | |
| Cation Exchange | |
| Antibodix |
| 1.7 μm | |
| Non-porous | |
| Stainless Steel | |
| HPLC |
Antibodix is a registered trademark of Sepax Technologies