missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Antibiotics: Nystatin

Serve variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis

$395.61 - $395.61

Specifications

Content And Storage 4°C
Form Powder
Product Type Antifungal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19453425
View Documents
MP Biomedicals™
0219453425
25 mU
Each for $395.61
Only null left
 
Description

Description

  • Format: Powder, Non-sterile

All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.

Specifications

Specifications

4°C
Powder
Antifungal
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'