missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Antibiotics: Nystatin
Serve variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis
Supplier: MP Biomedicals™ 0219453425
Description
- Format: Powder, Non-sterile
All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.
Specifications
| 4°C | |
| Powder | |
| Antifungal | |
| 25 mU |