missing translation for 'onlineSavingsMsg'
Learn More

unmethylated-Histone H3 (Lys9) Mouse anti-Human, Clone: 9B1-2G6, MilliporeSigma™

Mouse Monoclonal Antibody

$212.53 - $908.54

Specifications

Antigen unmethylated-Histone H3 (Lys9)
Clone 9B1-2G6
Applications Dot Blot, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABE263SMI
View Documents Promotion Details
N/A 10 μg
Each for $212.53
Only null left
 
MABE263MI
View Documents Promotion Details
N/A 100 μg
Each for $908.54
Only null left
 
Description

Description

Use Anti-unmethylated-Histone H3 (Lys9) Antibody, clone 9B1-2G6 (Mouse Monoclonal Antibody) demonstrated to detect unmethylated-Histone H3 (Lys9) also known as Histone H3.1.
Specifications

Specifications

unmethylated-Histone H3 (Lys9)
Dot Blot, Western Blot
Unconjugated
Mouse
Purified mouse monoclonal IgG1κ supernatant in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Hist1H3a, H3a, Hist1h3g, H3.1-221, H3g, Hist1h3h, H3.1-291, H3h, Hist1h3i, H3.1-l, H3i
IgG1 κ
Protein G purified
9B1-2G6
Monoclonal
Purified
Human
NP_038578
KLH-conjugated linear peptide corresponding to human unmethylated Histone H3 (Lys9).
Primary
2-8°C for one year
Videos
SDS
Documents

Documents

For Research Use Only