missing translation for 'onlineSavingsMsg'
Learn More
Learn More
unmethylated-Histone H3 (Lys9) Mouse anti-Human, Clone: 9B1-2G6, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABE263
Description
Use Anti-unmethylated-Histone H3 (Lys9) Antibody, clone 9B1-2G6 (Mouse Monoclonal Antibody) demonstrated to detect unmethylated-Histone H3 (Lys9) also known as Histone H3.1.Chemical Identifiers
| unmethylated-Histone H3 (Lys9) | |
| Dot Blot, Western Blot | |
| Unconjugated | |
| Mouse | |
| Purified mouse monoclonal IgG1κ supernatant in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| Hist1H3a, H3a, Hist1h3g, H3.1-221, H3g, Hist1h3h, H3.1-291, H3h, Hist1h3i, H3.1-l, H3i | |
| IgG1 κ | |
| Protein G purified |
| 9B1-2G6 | |
| Monoclonal | |
| Purified | |
| Human | |
| NP_038578 | |
| KLH-conjugated linear peptide corresponding to human unmethylated Histone H3 (Lys9). | |
| Primary | |
| 2-8°C for one year |
Specifications
| unmethylated-Histone H3 (Lys9) | |
| Monoclonal | |
| Unconjugated | |
| NP_038578 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| 2-8°C for one year | |
| IgG1 κ |
| Dot Blot, Western Blot | |
| 9B1-2G6 | |
| Purified mouse monoclonal IgG1κ supernatant in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| Hist1H3a, H3a, Hist1h3g, H3.1-221, H3g, Hist1h3h, H3.1-291, H3h, Hist1h3i, H3.1-l, H3i | |
| KLH-conjugated linear peptide corresponding to human unmethylated Histone H3 (Lys9). | |
| 100 μg | |
| Human | |
| Purified |
For Research Use Only