missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl-Histone H3 (Lys9) Rabbit anti-Human, Mouse, Chicken, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$164.48 - $754.51

Specifications

Antigen monomethyl-Histone H3 (Lys9)
Classification Polyclonal
Conjugate Unconjugated
Form Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
07450SMI
View Documents Promotion Details
N/A 10 μg
Each for $164.48
Only null left
 
07450MI
View Documents Promotion Details
N/A 100 μg
Each for $754.51
Only null left
 
Description

Description

Anti-monomethyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of Histone H3 monomethylated on lysine 9 (H3K9me1). This purified antibody is specificity verified by dot blot (DB), published in peer reviewed journals, and has been validated in immunocytochemistry, PIA and western blotting.
Specifications

Specifications

monomethyl-Histone H3 (Lys9)
Unconjugated
Rabbit
Purified rabbit IgG in buffer containing 0.1M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide and 30% glycerol.
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
IgG
Protein A purfied
Polyclonal
Purified
Chicken, Human, Mouse
NM_003493
KLH-conjugated, synthetic 2X-branched peptide containing the sequence ...ARmeKST... in which meK corresponds to monomethyl-lysine at residue 9 of human histone H3.
Primary
-20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Videos
SDS
Documents

Documents

For Research Use Only