Learn More
Monomethyl-Histone H3 (Lys9) Rabbit anti-Human, Mouse, Chicken, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier:  MilliporeSigma 07450S
 View more versions of this product
Catalog No. 07450SMI
Only null left
Description
Description
Anti-monomethyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of Histone H3 monomethylated on lysine 9 (H3K9me1). This purified antibody is specificity verified by dot blot (DB), published in peer reviewed journals, and has been validated in immunocytochemistry, PIA and western blotting.
Specifications
Chemical Identifiers
monomethyl-Histone H3 (Lys9)
Unconjugated
Rabbit
Purified rabbit IgG in buffer containing 0.1M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide and 30% glycerol.
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
IgG
Protein A purfied
Polyclonal
Purified
Chicken, Human, Mouse
NM_003493
KLH-conjugated, synthetic 2X-branched peptide containing the sequence ...ARmeKST... in which meK corresponds to monomethyl-lysine at residue 9 of human histone H3.
Primary
-20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Specifications
monomethyl-Histone H3 (Lys9)
Polyclonal
Purified rabbit IgG in buffer containing 0.1M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide and 30% glycerol.
Rabbit
Protein A purfied
Primary
Chicken, Human, Mouse
Purified
Immunocytochemistry, Inhibition Assays, Western Blot
Unconjugated
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
KLH-conjugated, synthetic 2X-branched peptide containing the sequence ...ARmeKST... in which meK corresponds to monomethyl-lysine at residue 9 of human histone H3.
10 μg
NM_003493
-20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
IgG
Product Suggestions
Product Suggestions