missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Histone H3 (Lys9) Mouse anti-Human, Vertebrates, Clone: CMA307, MilliporeSigma™
Mouse Monoclonal Antibody
$197.60 - $825.15
Specifications
| Antigen | Dimethyl Histone H3 (Lys9) |
|---|---|
| Clone | CMA307 |
| Applications | ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
05-124-9SMI
|
MilliporeSigma
051249S |
10 μg |
Each for $197.60
|
|
|||||
|
05-124-9MI
|
MilliporeSigma
MIL051249 |
100 μg |
Each for $825.15
|
|
|||||
Description
Use Anti-Dimethyl Histone H3 (Lys9) Antibody, clone CMA307 (mouse monoclonal antibody) validated in western blot, ELISA, immunocytochemistry, immunoprecipitation, Mplex to detect Dimethyl Histone H3 (Lys9) also known as H3K9me2, Histone H3 (di methyl K9).Specifications
| Dimethyl Histone H3 (Lys9) | |
| ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Mouse monoclonal IgG1κ supernatant in PBS with 0.05% sodium azide. | |
| H3.4, H3/g , H3/t , H3FT , H3T, H3t , MGC126886, MGC126888 , OTTHUMP00000037945 | |
| IgG1 κ | |
| Protein G purfied |
| CMA307 | |
| Monoclonal | |
| Purified | |
| Human, Vertebrates | |
| NM_003493 | |
| Synthetic peptide corresponding to amino acids 1-19 of human Histone H3, dimethylated on Lys9, conjugated to KLH. | |
| Primary | |
| 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. |
For Research Use Only