missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Histone H3 (Lys9) Mouse anti-Human, Vertebrates, Clone: CMA307, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051249S
Description
Use Anti-Dimethyl Histone H3 (Lys9) Antibody, clone CMA307 (mouse monoclonal antibody) validated in western blot, ELISA, immunocytochemistry, immunoprecipitation, Mplex to detect Dimethyl Histone H3 (Lys9) also known as H3K9me2, Histone H3 (di methyl K9).Specifications
| Dimethyl Histone H3 (Lys9) | |
| Monoclonal | |
| Unconjugated | |
| NM_003493 | |
| Mouse | |
| Protein G purfied | |
| Primary | |
| 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
| IgG1 κ |
| ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| CMA307 | |
| Mouse monoclonal IgG1κ supernatant in PBS with 0.05% sodium azide. | |
| H3.4, H3/g , H3/t , H3FT , H3T, H3t , MGC126886, MGC126888 , OTTHUMP00000037945 | |
| Synthetic peptide corresponding to amino acids 1-19 of human Histone H3, dimethylated on Lys9, conjugated to KLH. | |
| 10 μg | |
| Human, Vertebrates | |
| Purified |
For Research Use Only