missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Histone H3 (Arg26), Asymmetric Rabbit anti-Human, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$191.09 - $727.26

Specifications

Antigen dimethyl Histone H3 (Arg26), Asymmetric
Applications Dot Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ABE411SMI
View Documents
MilliporeSigma
ABE411S
10 μg
Each for $191.09
Only null left
 
ABE411MI
View Documents
MilliporeSigma
ABE411
100 μg
Each for $727.26
Only null left
 
Description

Description

This Anti-dimethyl Histone H3 (Arg26) antibody, Asymmetric is validated for use in western blotting, immunocytochemistry and dot blot for the detection of dimethyl Histone H3 (Arg26).
Specifications

Specifications

dimethyl Histone H3 (Arg26), Asymmetric
Polyclonal
Rabbit
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ
Primary
2-8°C for 12 months
Dot Blot, Immunocytochemistry, Western Blot
Unconjugated
Human
NP_003520
KLH-conjugated linear peptide corresponding to a region near the N-terminus of human Histone H3, asymmetrically dimethylated at Arg26.
Affinity Purified
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only