Learn More
Dimethyl Histone H3 (Arg26), Asymmetric Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier:  MilliporeSigma ABE411
 View more versions of this product
Catalog No. ABE411MI
Only null left
Explore available promotions
Description
Description
This Anti-dimethyl Histone H3 (Arg26) antibody, Asymmetric is validated for use in western blotting, immunocytochemistry and dot blot for the detection of dimethyl Histone H3 (Arg26).
Specifications
Specifications
dimethyl Histone H3 (Arg26), Asymmetric
Polyclonal
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Rabbit
Affinity Purified
Primary
Human
Dot Blot, Immunocytochemistry, Western Blot
Unconjugated
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ
KLH-conjugated linear peptide corresponding to a region near the N-terminus of human Histone H3, asymmetrically dimethylated at Arg26.
100 μg
NP_003520
2-8°C for 12 months
Product Suggestions
Product Suggestions