missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anthrone, 95%, pure
CAS: 90-44-8 | C14H10O | 194.23 g/mol
Supplier: Thermo Scientific Chemicals 401361000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Anthrone | |
| 152.0°C to 157.0°C | |
| 94% min. (GC) | |
| C14H10O | |
| 100 g | |
| 15,682 | |
| Solubility in water: insoluble. Other solubilities: soluble in hot naoh solution, conc. sulfuric acid, soluble in alcohol, benzene, aceton, soluble in most organic solvents without, fluorescence | |
| O=C1C2=CC=CC=C2CC2=CC=CC=C12 | |
| 194.23 | |
| CHEBI:33835 | |
| 95% |
| 90-44-8 | |
| 721.0°C | |
| Glass Bottle | |
| MFCD00001187 | |
| 07,473 | |
| anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744 | |
| RJGDLRCDCYRQOQ-UHFFFAOYSA-N | |
| 10H-anthracen-9-one | |
| 7018 | |
| 194.23 | |
| Pure |
Chemical Identifiers
| 90-44-8 | |
| 194.23 | |
| RJGDLRCDCYRQOQ-UHFFFAOYSA-N | |
| 7018 | |
| 10H-anthracen-9-one |
| C14H10O | |
| MFCD00001187 | |
| anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744 | |
| CHEBI:33835 | |
| O=C1C2=CC=CC=C2CC2=CC=CC=C12 |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 201-994-
RUO – Research Use Only