missing translation for 'onlineSavingsMsg'
Learn More

Anthrone, 95%, pure

Catalog No. p-4813328
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC401360250 25 g Glass bottle
AC401361000 100 g Glass bottle
AC401365000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC401360250 Supplier Thermo Scientific Chemicals Supplier No. 401360250
Only null left

CAS: 90-44-8 | C14H10O | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90-44-8
Molecular Formula C14H10O
Molecular Weight (g/mol) 194.23
MDL Number MFCD00001187
InChI Key RJGDLRCDCYRQOQ-UHFFFAOYSA-N
Synonym anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744
PubChem CID 7018
ChEBI CHEBI:33835
IUPAC Name 10H-anthracen-9-one
SMILES O=C1C2=CC=CC=C2CC2=CC=CC=C12

Specifications

Melting Point 152.0°C to 157.0°C
Boiling Point 721.0°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 07,473
Merck Index 15,682
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hot naoh solution, conc. sulfuric acid, soluble in alcohol, benzene, aceton, soluble in most organic solvents without, fluorescence
Formula Weight 194.23
Percent Purity 95%
Grade Pure
Chemical Name or Material Anthrone
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 201-994-

RUO – Research Use Only