missing translation for 'onlineSavingsMsg'
Learn More

Anthraquinone-2-carboxylic Acid 99.0+%, TCI America™

Catalog No. A05045G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A05045G 5 g
A050425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A05045G Supplier TCI America Supplier No. A05045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 117-78-2
Molecular Formula C15H8O4
Molecular Weight (g/mol) 252.225
MDL Number MFCD00001231
InChI Key ASDLSKCKYGVMAI-UHFFFAOYSA-N
Synonym anthraquinone-2-carboxylic acid, 9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid, 2-carboxyanthraquinone, 2-anthraquinonecarboxylic acid, 2-carboxyl-9,10-anthraquinone, anthraquinone-2-carboxylate, 9,10-dihydro-9,10-dioxo-2-anthracenecarboxylic acid, 9,10-anthraquinone 2-carboxylic acid, .beta.-anthraquinonecarboxylic acid, anthraquinone-.beta.-carboxylic acid
PubChem CID 67030
IUPAC Name 9,10-dioxoanthracene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)C(=O)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 252.23
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Anthraquinone-2-carboxylic Acid
EINECSNumber (4)-0723
TSCA No