accessibility menu, dialog, popup

UserName

CAS RN 117-78-2

IUPAC Name: 9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid
Synonyms: 9,10-dioxoanthracene-2-carboxylic acid
Molecular Weight (g/mol): 252.23
Molecular Formula: C15H8O4
InChi Key: ASDLSKCKYGVMAI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)