missing translation for 'onlineSavingsMsg'
Learn More

Anthracene 97.0+%, TCI America™

Supplier:  TCI America A0495500G

 View more versions of this product

Catalog No. A0495500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Anthracene
218°C
C14H10
500 g
paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767
C1=CC2=CC3=CC=CC=C3C=C2C=C1
178.23
CHEBI:35298
≥97.0% (GC)
120-12-7
342°C
MFCD00001240
3077
MWPLVEDNUUSJAV-UHFFFAOYSA-N
anthracene
8418
178.23
Crystalline Powder

Chemical Identifiers

120-12-7
178.23
MWPLVEDNUUSJAV-UHFFFAOYSA-N
8418
anthracene
C14H10
MFCD00001240
paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767
CHEBI:35298
C1=CC2=CC3=CC=CC=C3C=C2C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0683

RTECSNumber : CA9350000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions