missing translation for 'onlineSavingsMsg'
Learn More

Anthracene, 99%

CAS: 120-12-7 | C14H10 | 178.23 g/mol

Supplier:  Thermo Scientific Chemicals 104861000

 View more versions of this product

Catalog No. AC104861000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Anthracene
120-12-7
Yellow
121°C
98.5% min. (GC)
C14H10
100 g
12,290; 15,99; 16,13
paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767
MWPLVEDNUUSJAV-UHFFFAOYSA-N
anthracene
8418
178.23
Crystalline Powder
99%
215.0°C to 218.0°C
340.0°C
Authentic
Plastic bottle
MFCD00001240
05,657
15,674
Solubility in water: insoluble. Other solubilities: 1g/67mL alcohol - 1g/70mL methanol, 1g/200mL ether - 1g/62mL benzene, 1g/85mL chloroform - 1g/31mL carbon disulfide, 1g/86mL ccl4 - 1g/125mL toluene
C1=CC2=CC3=CC=CC=C3C=C2C=C1
178.23
CHEBI:35298
99%

Chemical Identifiers

120-12-7
178.23
MWPLVEDNUUSJAV-UHFFFAOYSA-N
8418
anthracene
C14H10
MFCD00001240
paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767
CHEBI:35298
C1=CC2=CC3=CC=CC=C3C=C2C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Warning

EINECSNumber : 204-371-1

RTECSNumber : CA9350000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only