missing translation for 'onlineSavingsMsg'
Learn More

Anthracene-9,10-dicarboxaldehyde 98.0+%, TCI America™

Catalog No. p-7118009
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26641G 1 g
A26645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26641G Supplier TCI America Supplier No. A26641G

Chemical Identifiers

CAS 7044-91-9
Molecular Formula C16H10O2
Molecular Weight (g/mol) 234.25
MDL Number MFCD00045386
InChI Key SBRUFOSORMQHES-UHFFFAOYSA-N
Synonym 9,10-anthracenedicarboxaldehyde, anthracene-9,10-dicarboxaldehyde, anthracene-9,10-dialdehyde, 9,10-diformylanthracene, pubchem15118, 9,10-anthracenedicarbaldehyde, 9,10-anthracenedicarbaldehyde #, 9,10-anthracene dicarboxyaldehyde
PubChem CID 81500
ChEBI CHEBI:51295
IUPAC Name anthracene-9,10-dicarbaldehyde
SMILES O=CC1=C2C=CC=CC2=C(C=O)C2=CC=CC=C12

Specifications

Melting Point 244°C
Quantity 1 g
Formula Weight 234.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Anthracene-9,10-dicarboxaldehyde