CAS RN 7044-91-9
IUPAC Name:
anthracene-9,10-dicarbaldehyde
Synonyms:
anthracene-9,10-dialdehyde
Molecular Weight (g/mol):
234.25
Molecular Formula:
C16H10O2
InChi Key:
SBRUFOSORMQHES-UHFFFAOYSA-N
SMILES:
O=CC1=C2C=CC=CC2=C(C=O)C2=CC=CC=C12
1
–
1
of
1
results
1
Anthracene-9,10-dicarboxaldehyde 98.0+%, TCI America™
CAS: 7044-91-9 Molecular Formula: C16H10O2 Molecular Weight (g/mol): 234.25 MDL Number: MFCD00045386 InChI Key: SBRUFOSORMQHES-UHFFFAOYSA-N Synonym: 9,10-anthracenedicarboxaldehyde,anthracene-9,10-dicarboxaldehyde,anthracene-9,10-dialdehyde,9,10-diformylanthracene,pubchem15118,9,10-anthracenedicarbaldehyde,9,10-anthracenedicarbaldehyde #,9,10-anthracene dicarboxyaldehyde PubChem CID: 81500 ChEBI: CHEBI:51295 IUPAC Name: anthracene-9,10-dicarbaldehyde SMILES: O=CC1=C2C=CC=CC2=C(C=O)C2=CC=CC=C12