Learn More
Anisole, 99%, Extra Dry, AcroSeal™
CAS: 100-66-3 | C7H8O | 108.14 g/mol
Supplier: Thermo Scientific Chemicals 429250010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Anisole | |
| -37.0°C | |
| 154.0°C | |
| 98.5% min. (GC) | |
| 1.5160 to 1.5190 | |
| 1 L | |
| 15,662 | |
| Solubility in water: 1.6g/L (20°C). Other solubilities: soluble in alcohol, ether, acetone | |
| COC1=CC=CC=C1 | |
| 108.14 | |
| CHEBI:16579 | |
| 99% |
| 100-66-3 | |
| 0.9900g/mL | |
| 43°C | |
| C7H8O | |
| C6H5OCH3 | |
| 0.99 | |
| methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene | |
| RDOXTESZEPMUJZ-UHFFFAOYSA-N | |
| anisole | |
| 7519 | |
| 108.14 | |
| Extra Dry |
Chemical Identifiers
| 100-66-3 | |
| 108.14 | |
| methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene | |
| CHEBI:16579 | |
| COC1=CC=CC=C1 |
| C7H8O | |
| RDOXTESZEPMUJZ-UHFFFAOYSA-N | |
| 7519 | |
| anisole |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ven
GHS Signal Word: Warning
EINECSNumber : 202-876-1
RUO – Research Use Only