missing translation for 'onlineSavingsMsg'
Learn More

Anisole, 99%, Extra Dry, AcroSeal™

Catalog No. p-4813326
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC429251000 100 mL
AC429250010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC429251000 Supplier Thermo Scientific Chemicals Supplier No. 429251000
Only null left

CAS: 100-66-3 | C7H8O | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-66-3
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
InChI Key RDOXTESZEPMUJZ-UHFFFAOYSA-N
Synonym methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene
PubChem CID 7519
ChEBI CHEBI:16579
IUPAC Name anisole
SMILES COC1=CC=CC=C1

Specifications

Melting Point -37.0°C
Density 0.9900g/mL
Boiling Point 154.0°C
Flash Point 43°C
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass bottle
Refractive Index 1.5160 to 1.5190
Linear Formula C6H5OCH3
Quantity 100 mL
Merck Index 15,662
Specific Gravity 0.99
Solubility Information Solubility in water: 1.6g/L (20°C). Other solubilities: soluble in alcohol, ether, acetone
Formula Weight 108.14
Percent Purity 99%
Grade Extra Dry
Chemical Name or Material Anisole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ven
EINECSNumber 202-876-1

RUO – Research Use Only