missing translation for 'onlineSavingsMsg'
Learn More

Anions - Sandy Loam 1, MilliporeSigma™ Supelco™

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

$352.85 - $391.69

Specifications

CAS 14808-60-7
Type Certified Reference Material
Grade Certified Reference Material
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111001634
View Documents
Merck Emd Millipore
CRM70030G
30 g
Each for $352.85
Only null left
 
111001630
View Documents
Merck Emd Millipore
CRM70050G
50 g
Each for $391.69
Only null left
 
Description

Description

Featured Industries: Environmental
Specifications

Specifications

14808-60-7
Certified Reference Material
Certified Reference Material
Videos
SDS
Documents

Documents