missing translation for 'onlineSavingsMsg'
Learn More

Anions - Sandy Loam 1, MilliporeSigma™ Supelco™

Catalog No. 111001634
Change view
Click to view available options
Quantity:
30 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001634 30 g
111001630 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111001634 Supplier Merck Emd Millipore Supplier No. CRM70030G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Featured Industries: Environmental

Specifications

CAS 14808-60-7
Type Certified Reference Material
Quantity 30 g
Grade Certified Reference Material