missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amyl Sulfide 97.0+%, TCI America™
$190.59 - $573.70
Chemical Identifiers
| CAS | 872-10-6 |
|---|---|
| Molecular Formula | C10H22S |
| Molecular Weight (g/mol) | 174.35 |
| MDL Number | MFCD00027275 |
| InChI Key | JOZDADPMWLVEJK-UHFFFAOYSA-N |
| Synonym | pentyl sulfide, diamyl sulfide, pentane, 1,1'-thiobis, amyl sulfide, n-amyl sulfide, dipentyl sulphide, di-n-pentyl sulfide, dipentyl sulfide, unii-61xjn3644r, di-n-pentyl sulphide |
| PubChem CID | 13382 |
| IUPAC Name | 1-(pentylsulfanyl)pentane |
| SMILES | CCCCCSCCCCC |
Chemical Identifiers
| 872-10-6 | |
| 174.35 | |
| JOZDADPMWLVEJK-UHFFFAOYSA-N | |
| 13382 | |
| CCCCCSCCCCC |
| C10H22S | |
| MFCD00027275 | |
| pentyl sulfide, diamyl sulfide, pentane, 1,1'-thiobis, amyl sulfide, n-amyl sulfide, dipentyl sulphide, di-n-pentyl sulfide, dipentyl sulfide, unii-61xjn3644r, di-n-pentyl sulphide | |
| 1-(pentylsulfanyl)pentane |
Specifications
| 872-10-6 | |
| 175°C | |
| MFCD00027275 | |
| pentyl sulfide, diamyl sulfide, pentane, 1,1'-thiobis, amyl sulfide, n-amyl sulfide, dipentyl sulphide, di-n-pentyl sulfide, dipentyl sulfide, unii-61xjn3644r, di-n-pentyl sulphide | |
| CCCCCSCCCCC | |
| 174.35 | |
| 174.35 | |
| Liquid |
| Colorless | |
| C10H22S | |
| 5 g | |
| JOZDADPMWLVEJK-UHFFFAOYSA-N | |
| 1-(pentylsulfanyl)pentane | |
| 13382 | |
| ≥97.0% (GC) | |
| Amyl Sulfide |
Safety and Handling
EINECSNumber : (2)-0472
TSCA : No