missing translation for 'onlineSavingsMsg'
Learn More

Amyl Benzoate 98.0+%, TCI America™

Catalog No. p-7118848
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B109525ML 25 mL
B1095100ML 100 mL
B1095500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B109525ML Supplier TCI America Supplier No. B109525ML

Chemical Identifiers

CAS 2049-96-9
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00048843
InChI Key QKNZNUNCDJZTCH-UHFFFAOYSA-N
Synonym amyl benzoate, n-pentyl benzoate, benzoic acid, pentyl ester, n-amyl benzoate, unii-cle7k4b29s, cle7k4b29s, amylbenzoat, amylbenzoate, n amyl benxoate, benzoic acid pentyl
PubChem CID 16296
IUPAC Name pentyl benzoate
SMILES CCCCCOC(=O)C1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 260°C
Quantity 25 mL
Formula Weight 192.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Amyl Benzoate
EINECSNumber (3)-1356
RTECSNumber DH6276000
TSCA Yes