missing translation for 'onlineSavingsMsg'
Learn More

Amyl Acetate 99.0+%, TCI America™

Supplier:  TCI America A0021500ML

 View more versions of this product

Catalog No. A0021500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Amyl Acetate
Colorless
C7H14O2
500 mL
amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether
CCCCCOC(=O)C
130.187
CHEBI:87362
≥99.0% (GC)
628-63-7
148°C
MFCD00009500
1104
PGMYKACGEOXYJE-UHFFFAOYSA-N
pentyl acetate
12348
130.19
Liquid

Chemical Identifiers

628-63-7
130.187
PGMYKACGEOXYJE-UHFFFAOYSA-N
12348
pentyl acetate
C7H14O2
MFCD00009500
amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether
CHEBI:87362
CCCCCOC(=O)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0733

RTECSNumber : AJ1925000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions