accessibility menu, dialog, popup

UserName

CAS RN 628-63-7

IUPAC Name: pentyl acetate
Synonyms: amyl acetate N-amyl acetate
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: PGMYKACGEOXYJE-UHFFFAOYSA-N
SMILES: CCCCCOC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (5)
  • (9)
Percent Purity 
  • (1)
  • (2)
  • (6)
  • (5)
Grade 
  • (1)
  • (1)
  • (6)
  • (3)
Boiling Point 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (1)
  • (1)
  • (9)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (5)
  • (9)
  • (1)
  • (2)
  • (6)
  • (5)
  • (1)
  • (1)
  • (6)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)