missing translation for 'onlineSavingsMsg'
Learn More

Amyl Acetate 99.0+%, TCI America™

Catalog No. p-7116662
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
A002125ML 25 mL
A0021500ML 500 mL
2 options

Catalog No. A002125ML

Supplier: TCI America A002125ML

Only null left

Chemical Identifiers

CAS 628-63-7
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009500
InChI Key PGMYKACGEOXYJE-UHFFFAOYSA-N
Synonym amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether
PubChem CID 12348
ChEBI CHEBI:87362
IUPAC Name pentyl acetate
SMILES CCCCCOC(=O)C

Specifications

Color Colorless
Boiling Point 148°C
Quantity 25 mL
UN Number 1104
Formula Weight 130.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Amyl Acetate
EINECSNumber (2)-0733
RTECSNumber AJ1925000
TSCA Yes