missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals AMPSO, 0.2M buffer soln., pH 9.0

Catalog No. aaj62328ak
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62328AK 250 mL
AAJ62328AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62328AK Supplier Thermo Scientific Chemicals Supplier No. J62328AK
Only null left

CAS: 102029-60-7 | C7H16NNaO5S | 249.257 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102029-60-7
Molecular Formula C7H16NNaO5S
Molecular Weight (g/mol) 249.257
MDL Number MFCD00067500
InChI Key UVGYCHPTWHWEPZ-UHFFFAOYSA-M
PubChem CID 44119996
IUPAC Name sodium;2-hydroxy-3-[(1-hydroxy-2-methylpropan-2-yl)amino]propane-1-sulfonate
SMILES CC(C)(CO)NCC(CS(=O)(=O)[O-])O.[Na+]

Specifications

Color Colorless
Concentration 0.2 M
pH 9.0
Physical Form Liquid
Quantity 250 mL
Odor Odorless
Chemical Name or Material AMPSO buffer soln.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only