missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Amprenavir

Catalog No. AC466980250
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466980250 25 mg
AC466981000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466980250 Supplier Thermo Scientific Chemicals Supplier No. 466980250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 161814-49-9 | C25H35N3O6S, C25H35N3O6S | 505.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 161814-49-9
Molecular Formula C25H35N3O6S

Specifications

Quantity 25 mg
Formula Weight 505.63
InChI Key YMARZQAQMVYCKC-OEMFJLHTSA-N
SMILES CC(C)CN(C[C@@H](O)[C@H](CC1=CC=CC=C1)NC(=O)O[C@H]1CCOC1)S(=O)(=O)C1=CC=C(N)C=C1
IUPAC Name (3S)-oxolan-3-yl N-[(2S,3R)-3-hydroxy-4-[N-(2-methylpropyl)4-aminobenzenesulfonamido]-1-phenylbutan-2-yl]carbamate
Molecular Weight (g/mol) 505.63
Chemical Name or Material Amprenavir

RUO – Research Use Only