missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin Trihydrate 98.0+%, TCI America™

Catalog No. p-7117725
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
A20925G 5 g
A209225G 25 g
2 options

Catalog No. A20925G

Supplier: TCI America A20925G

Only null left

Chemical Identifiers

CAS 7177-48-2
Molecular Formula C16H25N3O7S
Molecular Weight (g/mol) 403.45
MDL Number MFCD00072036
InChI Key RXDALBZNGVATNY-CWLIKTDRSA-N
Synonym ampicillin trihydrate, aminobenzylpenicillin trihydrate, amperil, ampinova, trafarbiot, amcap, cymbi, ampicillin 3-wasser, unii-hxq6a1n7r6, alpha-aminobenzylpenicillin trihydrate
PubChem CID 23565
ChEBI CHEBI:31209
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;trihydrate
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.O.O.O

Specifications

Melting Point 202°C
Color White-Yellow
Quantity 5 g
Formula Weight 349.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ampicillin Trihydrate
RTECSNumber XH8425000
TSCA No
Recommended Storage Refrigerator