missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Amphotericin B

Catalog No. 6930100
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6930100 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6930100 Supplier Tocris Bioscience™ Supplier No. 6930/100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Forms monovalent ion channels in lipid bilyers

Amphotericin B forms nonselective monovalent ion channels within lipid bilayers. Amphotericin B binds ergosterol in the cell membrane. Antifungal and antiprotozoal.

For more information about how Amphotericin B may be used, see our protocol:

3D Culture of Lung Alveolar Cells

Specifications

CAS 1397-89-3
Quantity 100 mg
Solubility Soluble to 20 mM in DMSO
Molecular Formula C47H73NO17
Formula Weight 924.09
Product Type Antifungal
Purity 90%