missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPD, 0.2M buffer soln., pH 9.5, Thermo Scientificâ„¢
$116.34 - $157.82
Specifications
CAS | 115-69-5 |
---|---|
Color | Colorless |
Concentration | 0.2 M |
pH | 9.5 |
Physical Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAJ60916AE
|
Thermo Scientific Chemicals
J60916AE |
100 mL |
Each for $116.34
|
|
||||||
AAJ60916AK
|
Thermo Scientific Chemicals
J60916AK |
250 mL |
Each for $157.82
|
|
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
115-69-5 | |
0.2 M | |
Liquid | |
105.137 |
Colorless | |
9.5 | |
Odorless |