Learn More
Ampac™ Flexibles SealPAK Pouches
Ideal for histology and pathology specimens and for safe biological waste disposal in medical, institutional and industrial facilities
$130.33 - $1,513.07
Specifications
Temperature Range (Metric) -56°C to +115°C
Thickness (English) 2.50 mil
View More Specs
Products 8
Catalog Number Mfr. No. Dimensions (L x W) Capacity (English) Material Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Capacity (English) Material Quantity Price Quantity & Availability  
0181216
Accessories View Documents Promotion Details
Ampac™
402 24
8 x 6.5 in. (20.3 x 16.5cm) 16 oz. Polyester 1800/Cs.
Pack of 75 Each for $133.53
Case of 24 Pack for $3,204.72
Only null left
0181225AA
Accessories View Documents Promotion Details
Ampac™
KAP403 24
9.5 x 8 in. (24.1 x 20.3cm) 24 oz. Polyester 1200/Cs.
Pack of 50 Each for $149.90
Case of 24 Pack for $3,597.60
Only null left
0181225BB
Accessories View Documents Promotion Details
Ampac™
405 24
12 x 10 in. (30.5 x 25.4cm) 48 oz. Polyester 720/Cs.
Pack of 30 Each for $133.30
Case of 24 Pack for $3,199.20
Only null left
0181225GG
Accessories View Documents Promotion Details
Ampac™
408 24
16 x 12 in. (40.6 x 30.5cm) 96 oz. Polyester 360/Cs.
Pack of 15 Each for $149.17
Case of 24 Pack for $3,580.08
Only null left
01812780
View Documents Promotion Details
Ampac™
400F31MB
6 x 4 in. (15.2 x 10.2cm) 4 oz. Foil 2400/Cs.
Case of 1000 Each for $1,513.07
Only null left
0181217
Accessories View Documents Promotion Details
Ampac™
404 24
12 x 8 in. (30.5 x 20.3cm) 32 oz. Polyester 960/Cs.
Pack of 40 Each for $130.33
Case of 24 Pack for $3,127.92
Only null left
0181225A
Accessories View Documents Promotion Details
Ampac™
400 24
6 x 4 in. (15.2 x 10.2cm) 4 oz. PET, Polyethylene 2400/Cs.
Pack of 100 Each for $149.17
Case of 24 Pack for $3,580.08
Only null left
0181225B
Accessories View Documents Promotion Details
Ampac™
KAP406 24
16 x 9.5 in. (40.6 x 24.1cm) 64 oz. Polyester 480/Cs.
Pack of 20 Each for $141.63
Case of 24 Pack for $3,399.12
Only null left
Description
Description
Ideal for histology and pathology specimens and for safe biological waste disposal in medical, institutional and industrial facilities
Recommended for applications requiring long-term storage, sharper objects, or chemical solutions.
  • Transparent pouches with polyethylene lining
  • Can withstand infrared and gamma rays
  • May be frozen, microwaved, or boiled without harm
  • Easily resealable
  • 2.5 mil
  • Use with Ampac Flexibles pouch sealer (Cat. Nos. 01-812-15 or 01-812-15B) and handy pouch cutter (01-812-21)
Order Info
Also available in heat-sealable tubular roll stock that allows you to create custom-size pouches.
Specifications
Specifications
-56°C to +115°C
2.50 mil